missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%

Catalog No. aal1955603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1955603 1 g
AAL19556MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1955603 Supplier Thermo Scientific Chemicals Supplier No. L1955603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 732306-24-0 | C6H4Cl2F3N | 218.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 732306-24-0
Molecular Formula C6H4Cl2F3N
Molecular Weight (g/mol) 218.00
MDL Number MFCD04972770
InChI Key HCKXJUZITVCDND-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt
PubChem CID 2782815
IUPAC Name 4-chloro-3-(trifluoromethyl)pyridine hydrochloride
SMILES Cl.FC(F)(F)C1=C(Cl)C=CN=C1

Specifications

Melting Point ∼135°C (sublimation)
Quantity 1 g
Formula Weight 218.01
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only