missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate, 97%

Catalog No. AAL1012706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1012706 5 g
AAL1012703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1012706 Supplier Thermo Scientific Chemicals Supplier No. L1012706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23163-86-2 | C8H3ClF3NS | 237.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23163-86-2
Molecular Formula C8H3ClF3NS
Molecular Weight (g/mol) 237.624
MDL Number MFCD00041090
InChI Key AHFPRSSHNSGRCU-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenyl isothiocyanate, 1-chloro-4-isothiocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisothiocyanate, 4-chloro-3-trifluoromethylphenyl isothiocyanate, 1-chloro-4-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-chloro-4-isothiocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethyl benzenisothiocyanate, acmc-20amlf, pubchem5016, 4-chloro-3-trifluoromethylphenylisothiocyanate
PubChem CID 520026
IUPAC Name 1-chloro-4-isothiocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1N=C=S)C(F)(F)F)Cl

Specifications

Density 1.47
Boiling Point 248°C to 251°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.59
Quantity 5 g
UN Number UN2922
Beilstein 787371
Sensitivity Moisture sensitive
Formula Weight 237.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only