missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-(trifluoromethyl)phenyl Isocyanate 98.0+%, TCI America™
$130.48 - $415.68
Chemical Identifiers
| CAS | 327-78-6 |
|---|---|
| Molecular Formula | C8H3ClF3NO |
| Molecular Weight (g/mol) | 221.563 |
| MDL Number | MFCD00013874 |
| InChI Key | NBJZEUQTGLSUOB-UHFFFAOYSA-N |
| Synonym | 4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 |
| PubChem CID | 2733265 |
| IUPAC Name | 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene |
| SMILES | C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl |
Chemical Identifiers
| 327-78-6 | |
| 221.563 | |
| NBJZEUQTGLSUOB-UHFFFAOYSA-N | |
| 2733265 | |
| C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl |
| C8H3ClF3NO | |
| MFCD00013874 | |
| 4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 | |
| 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene |
Specifications
| 327-78-6 | |
| White-Yellow | |
| C8H3ClF3NO | |
| 5 g | |
| 4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 | |
| C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl | |
| 221.563 | |
| 221.56 | |
| Crystalline Lumps |
| 36°C | |
| 92°C | |
| MFCD00013874 | |
| 2206 | |
| NBJZEUQTGLSUOB-UHFFFAOYSA-N | |
| 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene | |
| 2733265 | |
| ≥98.0% (GC) | |
| 4-Chloro-3-(trifluoromethyl)phenyl Isocyanate |
Safety and Handling
TSCA : No