missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 98%

Catalog No. AC309130050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309130050 5 g Glass bottle
AC309130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC309130050 Supplier Thermo Scientific Chemicals Supplier No. 309130050

CAS: 327-78-6 | C8H3ClF3NO | 221.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 327-78-6
Molecular Formula C8H3ClF3NO
Molecular Weight (g/mol) 221.56
MDL Number MFCD00013874
InChI Key NBJZEUQTGLSUOB-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027
PubChem CID 2733265
IUPAC Name 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 37.0°C
Color White to Yellow
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(CF3)NCO
Quantity 5 g
Formula Weight 221.56
Percent Purity 98%
Physical Form Low Melting Solid, Powder or Chunks
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only