missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. p-7125357
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22465G 5 g
C224625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22465G Supplier TCI America Supplier No. C22465G

Chemical Identifiers

CAS 1735-54-2
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.564
MDL Number MFCD00190117
InChI Key VBPYHJRQZZCFCW-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzonitrile, 2-chloro-5-cyanobenzotrifluoride, 3-trifluoromethyl-4-chlorobenzonitrile, benzonitrile, 4-chloro-3-trifluoromethyl, 4-chloro-3-trifluoromethyl benzenecarbonitrile, pubchem8192, ksc494o5b, 4-chloro-3-trifluoromethyl-benzonitrile
PubChem CID 2736474
IUPAC Name 4-chloro-3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C#N)C(F)(F)F)Cl

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 105°C
Quantity 5 g
UN Number 3439
Formula Weight 205.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzonitrile