missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. C28031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28031G 1 g
C28035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28031G Supplier TCI America Supplier No. C28031G

Chemical Identifiers

CAS 1737-36-6
Molecular Formula C8H3ClF3O2
Molecular Weight (g/mol) 223.56
MDL Number MFCD01631468
InChI Key PPHHAZOVVZBSCM-UHFFFAOYSA-M
Synonym 4-chloro-3-trifluoromethyl benzoic acid, benzoic acid, 4-chloro-3-trifluoromethyl, 5-carboxy-2-chlorobenzotrifluoride, 4-chloro-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-chlorobenzoic acid, pubchem4588, intermediates-zcf02292, ksc496a8t, 4-chloro-3-trifluoromethyl
PubChem CID 2773857
IUPAC Name 4-chloro-3-(trifluoromethyl)benzoate
SMILES [O-]C(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F

Specifications

Melting Point 160°C
Color White
Quantity 1 g
Formula Weight 224.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzoic Acid