missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoic Acid 98.0+%, TCI America™

Catalog No. A1189500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1189500G 500 g
A118925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1189500G Supplier TCI America Supplier No. A1189500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1205-30-7
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00012375
InChI Key FHQAWINGVCDTTG-UHFFFAOYSA-N
Synonym 4-chloro-5-sulphamoylbenzoic acid, 3-aminosulfonyl-4-chlorobenzoic acid, csba, sulfamido-3-chlorobenzoic acid, benzoic acid, 3-aminosulfonyl-4-chloro, 3-sulfamoyl-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-sulfamoyl, 4-chloro-3-sulfamoyl-benzoic acid, 4-chloro-5-sulfamoylbenzoic acid, 4-chloro-3-sulphamoylbenzoic acid
PubChem CID 14568
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl

Specifications

Melting Point 261°C
Color White
Quantity 500 g
Formula Weight 235.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic Acid
RTECSNumber DG5642600
TSCA No