missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoic acid, 98%

CAS: 1205-30-7 | C7H6ClNO4S | 235.65 g/mol

$91.53 - $91.53

Chemical Identifiers

CAS 1205-30-7
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.65
InChI Key FHQAWINGVCDTTG-UHFFFAOYSA-N
Synonym 4-chloro-5-sulphamoylbenzoic acid, 3-aminosulfonyl-4-chlorobenzoic acid, csba, sulfamido-3-chlorobenzoic acid, benzoic acid, 3-aminosulfonyl-4-chloro, 3-sulfamoyl-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-sulfamoyl, 4-chloro-3-sulfamoyl-benzoic acid, 4-chloro-5-sulfamoylbenzoic acid, 4-chloro-3-sulphamoylbenzoic acid
PubChem CID 14568
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC303220010
SDS View Documents
Thermo Scientific Chemicals
303220010
1 g
Each for $91.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1205-30-7
235.65
4-chloro-5-sulphamoylbenzoic acid, 3-aminosulfonyl-4-chlorobenzoic acid, csba, sulfamido-3-chlorobenzoic acid, benzoic acid, 3-aminosulfonyl-4-chloro, 3-sulfamoyl-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-sulfamoyl, 4-chloro-3-sulfamoyl-benzoic acid, 4-chloro-5-sulfamoylbenzoic acid, 4-chloro-3-sulphamoylbenzoic acid
4-chloro-3-sulfamoylbenzoic acid
C7H6ClNO4S
FHQAWINGVCDTTG-UHFFFAOYSA-N
14568
C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl

Specifications

1205-30-7
97.5% min. (HPLC)
C7H6ClNO4S
4-chloro-5-sulphamoylbenzoic acid, 3-aminosulfonyl-4-chlorobenzoic acid, csba, sulfamido-3-chlorobenzoic acid, benzoic acid, 3-aminosulfonyl-4-chloro, 3-sulfamoyl-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-sulfamoyl, 4-chloro-3-sulfamoyl-benzoic acid, 4-chloro-5-sulfamoylbenzoic acid, 4-chloro-3-sulphamoylbenzoic acid
C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl
235.65
235.65
4-Chloro-3-sulfamoylbenzoic acid
256.0°C to 258.0°C
Glass bottle
1 g
FHQAWINGVCDTTG-UHFFFAOYSA-N
4-chloro-3-sulfamoylbenzoic acid
14568
98%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 214-882-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only