missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoic acid, 98%

Catalog No. AC303220050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC303220050 5 g
AC303220010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303220050 Supplier Thermo Scientific Chemicals Supplier No. 303220050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1205-30-7 | C7H6ClNO4S | 235.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1205-30-7
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.65
InChI Key FHQAWINGVCDTTG-UHFFFAOYSA-N
Synonym 4-chloro-5-sulphamoylbenzoic acid, 3-aminosulfonyl-4-chlorobenzoic acid, csba, sulfamido-3-chlorobenzoic acid, benzoic acid, 3-aminosulfonyl-4-chloro, 3-sulfamoyl-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-sulfamoyl, 4-chloro-3-sulfamoyl-benzoic acid, 4-chloro-5-sulfamoylbenzoic acid, 4-chloro-3-sulphamoylbenzoic acid
PubChem CID 14568
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl

Specifications

Boiling Point 256.0°C to 258.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 235.65
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic acid
Hazard Category GHS Signal Word: Warning
EINECSNumber 214-882-1

RUO – Research Use Only