missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrotoluene 97.0+%, TCI America™

Catalog No. C023225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C023225G Supplier TCI America Supplier No. C023225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-60-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007085
InChI Key NWESJZZPAJGHRZ-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrotoluene, 3-nitro-4-chlorotoluene, benzene, 1-chloro-4-methyl-2-nitro, 2-chloro-5-methylnitrobenzene, 1-chloro-4-methyl-2-nitro-benzene, toluene, 4-chloro-3-nitro, 3-nitro-4-chlorotoluol, pubchem19752, 4,3-chloronitrotoluene, 4-chloro-3-nitro-toluene
PubChem CID 6976
IUPAC Name 1-chloro-4-methyl-2-nitrobenzene
SMILES CC1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Boiling Point 260°C
Quantity 25 g
UN Number 2433
Formula Weight 171.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-nitrotoluene
EINECSNumber (3)-0449
TSCA Yes