missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrophenol, 99%

Catalog No. AAL1010806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1010806 5 g
AAL1010814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1010806 Supplier Thermo Scientific Chemicals Supplier No. L1010806

CAS: 610-78-6 | C6H4ClNO3 | 173.552 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-78-6
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.552
MDL Number MFCD00043546
InChI Key JUIKCULGDIZNDI-UHFFFAOYSA-N
PubChem CID 69127
IUPAC Name 4-chloro-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])Cl

Specifications

Melting Point 124°C to 129°C
Odor Characteristic
Quantity 5 g
Beilstein 2094546
Synonym phenol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenol, 4chloro-3-nitrophenol, pubchem10459, 4-chlor-3-nitro-phenol, 3-nitro-4-chloro phenol, acmc-1azj5, phenol,4-chloro-3-nitro, 4-chloranyl-3-nitro-phenol, 2-chloro-5-hydroxy nitrobenzene
Formula Weight 173.56
Percent Purity 99%
Chemical Name or Material 4-Chloro-3-nitrophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only