missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrocinnamic Acid 97.0+%, TCI America™

Catalog No. p-7124852
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12055G 5 g
C120525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12055G Supplier TCI America Supplier No. C12055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20797-48-2
Molecular Formula C9H6ClNO4
Molecular Weight (g/mol) 227.6
MDL Number MFCD00063311
InChI Key QBDALTIMHOITIU-DUXPYHPUSA-N
Synonym 4-chloro-3-nitrocinnamic acid, trans-4-chloro-3-nitrocinnamic acid, 3-4-chloro-3-nitrophenyl acrylic acid, e-3-4-chloro-3-nitrophenyl acrylic acid, 2e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, 4-chloro-3-nitro-cinnamic acid, 3-nitro-4-chlorocinnamic acid, qbdaltimhoitiu-duxpyhpusa, 3-4-chloro-3-nitrophenyl propenoic acid
PubChem CID 688108
SMILES C1=CC(=C(C=C1C=CC(=O)O)[N+](=O)[O-])Cl

Specifications

Melting Point 187°C
Quantity 5 g
Formula Weight 227.60
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrocinnamic Acid