accessibility menu, dialog, popup

UserName

OOClNOO(E)

CAS RN 20797-48-2

IUPAC Name: (2E)-3-(4-chloro-3-nitrophenyl)prop-2-enoate
Synonyms: (2E)-3-(4-chloro-3-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol): 226.59
Molecular Formula: C9H5ClNO4
InChi Key: QBDALTIMHOITIU-DUXPYHPUSA-M
SMILES: [O-]C(=O)\C=C\C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)