missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrocinnamic acid, 98%

Catalog No. AAA1495406
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1495406 5 g
AAA1495422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1495406 Supplier Thermo Scientific Chemicals Supplier No. A1495406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20797-48-2 | C9H6ClNO4 | 227.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20797-48-2
Molecular Formula C9H6ClNO4
Molecular Weight (g/mol) 227.6
MDL Number MFCD00063311
InChI Key QBDALTIMHOITIU-DUXPYHPUSA-N
Synonym 4-chloro-3-nitrocinnamic acid, trans-4-chloro-3-nitrocinnamic acid, 3-4-chloro-3-nitrophenyl acrylic acid, e-3-4-chloro-3-nitrophenyl acrylic acid, 2e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, 4-chloro-3-nitro-cinnamic acid, 3-nitro-4-chlorocinnamic acid, qbdaltimhoitiu-duxpyhpusa, 3-4-chloro-3-nitrophenyl propenoic acid
PubChem CID 688108
SMILES C1=CC(=C(C=C1C=CC(=O)O)[N+](=O)[O-])Cl

Specifications

Melting Point 186°C to 188°C
Quantity 5 g
Formula Weight 227.61
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrocinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only