missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrobenzyl Alcohol 98.0+%, TCI America™
$123.03 - $384.00
Chemical Identifiers
| CAS | 55912-20-4 |
|---|---|
| Molecular Formula | C7H6ClNO3 |
| Molecular Weight (g/mol) | 187.579 |
| MDL Number | MFCD00007086 |
| InChI Key | QLLRQJDSYJIXTN-UHFFFAOYSA-N |
| PubChem CID | 91933 |
| IUPAC Name | (4-chloro-3-nitrophenyl)methanol |
| SMILES | C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl |
Chemical Identifiers
| 55912-20-4 | |
| 187.579 | |
| QLLRQJDSYJIXTN-UHFFFAOYSA-N | |
| (4-chloro-3-nitrophenyl)methanol |
| C7H6ClNO3 | |
| MFCD00007086 | |
| 91933 | |
| C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl |
Specifications
| 55912-20-4 | |
| C7H6ClNO3 | |
| 5 g | |
| QLLRQJDSYJIXTN-UHFFFAOYSA-N | |
| (4-chloro-3-nitrophenyl)methanol | |
| 91933 | |
| ≥98.0% (GC) | |
| 4-Chloro-3-nitrobenzyl Alcohol |
| 66°C | |
| MFCD00007086 | |
| 4-chloro-3-nitrobenzyl alcohol, 4-chloro-3-nitrophenyl methanol, 4-chloro-3-nitrobenzylalcohol, 4-chloro-3-nitrobenzenemethanol, 3-nitro-4-chlorobenzyl alcohol, acmc-1ayo8, qllrqjdsyjixtn-uhfffaoysa, benzenemethanol,4-chloro-3-nitro, benzenemethanol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenyl-methanol | |
| C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl | |
| 187.579 | |
| 187.58 | |
| Crystalline Powder |
Safety and Handling
TSCA : No