missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrobenzyl Alcohol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 66°C |
| Quantity | 5 g |
| Synonym | 4-chloro-3-nitrobenzyl alcohol, 4-chloro-3-nitrophenyl methanol, 4-chloro-3-nitrobenzylalcohol, 4-chloro-3-nitrobenzenemethanol, 3-nitro-4-chlorobenzyl alcohol, acmc-1ayo8, qllrqjdsyjixtn-uhfffaoysa, benzenemethanol,4-chloro-3-nitro, benzenemethanol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenyl-methanol |
| Formula Weight | 187.58 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-3-nitrobenzyl Alcohol |
Safety and Handling
| TSCA | No |