missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrobenzyl alcohol, 98%
CAS: 55912-20-4 | C7H6ClNO3 | 187.579 g/mol
$55.08 - $178.30
Chemical Identifiers
| CAS | 55912-20-4 |
|---|---|
| Molecular Formula | C7H6ClNO3 |
| Molecular Weight (g/mol) | 187.579 |
| MDL Number | MFCD00007086 |
| InChI Key | QLLRQJDSYJIXTN-UHFFFAOYSA-N |
| Synonym | 4-chloro-3-nitrobenzyl alcohol, 4-chloro-3-nitrophenyl methanol, 4-chloro-3-nitrobenzylalcohol, 4-chloro-3-nitrobenzenemethanol, 3-nitro-4-chlorobenzyl alcohol, acmc-1ayo8, qllrqjdsyjixtn-uhfffaoysa, benzenemethanol,4-chloro-3-nitro, benzenemethanol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenyl-methanol |
| PubChem CID | 91933 |
| IUPAC Name | (4-chloro-3-nitrophenyl)methanol |
| SMILES | C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1602406
|
Thermo Scientific Chemicals
A1602406 |
5 g |
Each for $55.08
|
|
|||||
|
AAA1602414
|
Thermo Scientific Chemicals
A1602414 |
25 g |
Each for $178.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 55912-20-4 | |
| 187.579 | |
| QLLRQJDSYJIXTN-UHFFFAOYSA-N | |
| 91933 | |
| C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl |
| C7H6ClNO3 | |
| MFCD00007086 | |
| 4-chloro-3-nitrobenzyl alcohol, 4-chloro-3-nitrophenyl methanol, 4-chloro-3-nitrobenzylalcohol, 4-chloro-3-nitrobenzenemethanol, 3-nitro-4-chlorobenzyl alcohol, acmc-1ayo8, qllrqjdsyjixtn-uhfffaoysa, benzenemethanol,4-chloro-3-nitro, benzenemethanol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenyl-methanol | |
| (4-chloro-3-nitrophenyl)methanol |
Specifications
| 55912-20-4 | |
| C7H6ClNO3 | |
| 5 g | |
| QLLRQJDSYJIXTN-UHFFFAOYSA-N | |
| (4-chloro-3-nitrophenyl)methanol | |
| 91933 | |
| 98% |
| 62°C to 64°C | |
| MFCD00007086 | |
| 4-chloro-3-nitrobenzyl alcohol, 4-chloro-3-nitrophenyl methanol, 4-chloro-3-nitrobenzylalcohol, 4-chloro-3-nitrobenzenemethanol, 3-nitro-4-chlorobenzyl alcohol, acmc-1ayo8, qllrqjdsyjixtn-uhfffaoysa, benzenemethanol,4-chloro-3-nitro, benzenemethanol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenyl-methanol | |
| C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl | |
| 187.579 | |
| 187.58 | |
| 4-Chloro-3-nitrobenzyl alcohol |
Safety and Handling
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 259-901-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only