missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzyl alcohol, 98%

Catalog No. AAA1602406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1602406 5 g
AAA1602414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1602406 Supplier Thermo Scientific Chemicals Supplier No. A1602406
Only null left

CAS: 55912-20-4 | C7H6ClNO3 | 187.579 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55912-20-4
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00007086
InChI Key QLLRQJDSYJIXTN-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzyl alcohol, 4-chloro-3-nitrophenyl methanol, 4-chloro-3-nitrobenzylalcohol, 4-chloro-3-nitrobenzenemethanol, 3-nitro-4-chlorobenzyl alcohol, acmc-1ayo8, qllrqjdsyjixtn-uhfffaoysa, benzenemethanol,4-chloro-3-nitro, benzenemethanol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenyl-methanol
PubChem CID 91933
IUPAC Name (4-chloro-3-nitrophenyl)methanol
SMILES C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl

Specifications

Melting Point 62°C to 64°C
Quantity 5 g
Formula Weight 187.58
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrobenzyl alcohol
Hazard Category H315-H317-H319-H335
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 259-901-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only