missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrobenzotrifluoride 97.0+%, TCI America™
$69.31 - $304.18
Chemical Identifiers
| CAS | 121-17-5 |
|---|---|
| Molecular Formula | C7H3ClF3NO2 |
| Molecular Weight (g/mol) | 225.55 |
| MDL Number | MFCD00007084 |
| InChI Key | TZGFQIXRVUHDLE-UHFFFAOYSA-N |
| Synonym | 4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride |
| PubChem CID | 8462 |
| IUPAC Name | 1-chloro-2-nitro-4-(trifluoromethyl)benzene |
| SMILES | [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F |
Chemical Identifiers
| 121-17-5 | |
| 225.55 | |
| TZGFQIXRVUHDLE-UHFFFAOYSA-N | |
| 8462 | |
| [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F |
| C7H3ClF3NO2 | |
| MFCD00007084 | |
| 4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride | |
| 1-chloro-2-nitro-4-(trifluoromethyl)benzene |
Specifications
| 121-17-5 | |
| 222°C | |
| MFCD00007084 | |
| 2307 | |
| TZGFQIXRVUHDLE-UHFFFAOYSA-N | |
| 1-chloro-2-nitro-4-(trifluoromethyl)benzene | |
| 8462 | |
| ≥97.0% (GC) | |
| 4-Chloro-3-nitrobenzotrifluoride |
| Yellow | |
| C7H3ClF3NO2 | |
| 25 g | |
| 4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride | |
| [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F | |
| 225.55 | |
| 225.55 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0452
RTECSNumber : XS9140000
TSCA : Yes