missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzotrifluoride 97.0+%, TCI America™

Catalog No. C0225500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0225500G 500 g
C022525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0225500G Supplier TCI America Supplier No. C0225500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-17-5
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.55
MDL Number MFCD00007084
InChI Key TZGFQIXRVUHDLE-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride
PubChem CID 8462
IUPAC Name 1-chloro-2-nitro-4-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F

Specifications

Color Yellow
Boiling Point 222°C
Quantity 500 g
UN Number 2307
Formula Weight 225.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-nitrobenzotrifluoride
EINECSNumber (3)-0452
RTECSNumber XS9140000
TSCA Yes