missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitro-2-pyridone, 98%

Catalog No. p-7046676
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1568606 5 g
AAL1568614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1568606 Supplier Thermo Scientific Chemicals Supplier No. L1568606
Only null left

CAS: 165547-79-5 | C5H3ClN2O3 | 174.54 g/mol

4-Chloro-3-nitro-2-pyridone finds an important application as raw material in organic synthesis and pharmaceuticals. It is also used as an intermediate in agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-3-nitro-2-pyridone finds an important application as raw material in organic synthesis and pharmaceuticals. It is also used as an intermediate in agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry condition. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 165547-79-5
Molecular Formula C5H3ClN2O3
Molecular Weight (g/mol) 174.54
MDL Number MFCD03093067
InChI Key UKIZCTHOMJXNIX-UHFFFAOYSA-N
Synonym 4-chloro-2-hydroxy-3-nitropyridine, 4-chloro-3-nitropyridin-2-ol, 4-chloro-3-nitropyridone, 4-chloro-3-nitro-2-pyridone, 4-chloro-3-nitropyridin-2 1h-one, 4-chloro-3-nitro-pyridin-2-ol, 4-chloro-3-nitro-pyridin-2-one, 2 1h-pyridinone, 4-chloro-3-nitro, pubchem6547, acmc-209dsr
PubChem CID 519282
IUPAC Name 4-chloro-3-nitro-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]

Specifications

Melting Point 222°C to 224°C
Quantity 5 g
Beilstein 149557
Solubility Information Insoluble in water.
Formula Weight 174.54
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitro-2-pyridone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only