CAS RN 165547-79-5
CAS RN 165547-79-5
IUPAC Name:
4-chloro-3-nitro-1,2-dihydropyridin-2-one
Synonyms:
4-chloro-3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol):
174.54
Molecular Formula:
C5H3ClN2O3
InChi Key:
UKIZCTHOMJXNIX-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=C(Cl)C=CNC1=O
1
–
1
of
1
results
1
4-Chloro-3-nitro-2-pyridone, 98%
CAS: 165547-79-5 Molecular Formula: C5H3ClN2O3 Molecular Weight (g/mol): 174.54 MDL Number: MFCD03093067 InChI Key: UKIZCTHOMJXNIX-UHFFFAOYSA-N Synonym: 4-chloro-2-hydroxy-3-nitropyridine,4-chloro-3-nitropyridin-2-ol,4-chloro-3-nitropyridone,4-chloro-3-nitro-2-pyridone,4-chloro-3-nitropyridin-2 1h-one,4-chloro-3-nitro-pyridin-2-ol,4-chloro-3-nitro-pyridin-2-one,2 1h-pyridinone, 4-chloro-3-nitro,pubchem6547,acmc-209dsr PubChem CID: 519282 IUPAC Name: 4-chloro-3-nitro-1H-pyridin-2-one SMILES: C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]