missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$96.94 - $96.94
Specifications
| CAS | 161950-10-3 |
|---|---|
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C7H8BClO2 |
| MDL Number | MFCD02683104 |
| InChI Key | UZDPQDBLCJDUAX-UHFFFAOYSA-N |
Specifications
| 161950-10-3 | |
| C7H8BClO2 | |
| UZDPQDBLCJDUAX-UHFFFAOYSA-N | |
| 3786632 |
| Crystal-Powder at 20°C | |
| MFCD02683104 | |
| (4-chloro-3-methylphenyl)boronic acid | |
| 170.40 |