missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylphenol, MilliporeSigma™ Supelco™

Catalog No. 111020080
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020080 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020080 Supplier Merck Emd Millipore Supplier No. 48519

4-Chloro-3-methylphenol is a biocide, which is colorless crystalline compound. It can be commonly used as a preservative in pharmaceutical products and as an additive to building materials and metal-working fluids.

Environmental

Specifications

Packaging Ampule
CAS 59-50-7
Flash Point 118°C
Physical Form Neat
Linear Formula ClC6H3(CH3)OH
MDL Number MFCD00002323
Melting Point 63°C to 65°C
Boiling Point 235°C
Synonym 4-Chloro-m-cresol; PCMC
Type Organic Pollutant Standard
Quantity 5000 mg
Formula Weight 142.58
Grade Analytical Standard
Show More Show Less