missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. C21155G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C21155G Supplier TCI America Supplier No. C21155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7697-29-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045853
InChI Key MRUKIIWRMSYKML-UHFFFAOYSA-N
Synonym 4-chloro-m-toluic acid, 4-chloro-3-methyl-benzoic acid, 4-chloro-3-methyl benzoic acid, benzoic acid, 4-chloro-3-methyl, m-toluic acid, 4-chloro, pubchem3666, 5-carboxy-2-chlorotoluene, acmc-1bg77, ksc497q5r, 4-chloranyl-3-methyl-benzoic acid
PubChem CID 282989
IUPAC Name 4-chloro-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1Cl)C(O)=O

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-methylbenzoic Acid