missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylbenzoic acid, 98%

Catalog No. AAL0853318
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0853318 50 g
AAL0853304 2 g
AAL0853309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0853318 Supplier Thermo Scientific Chemicals Supplier No. L0853318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7697-29-2 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7697-29-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045853
InChI Key MRUKIIWRMSYKML-UHFFFAOYSA-N
Synonym 4-chloro-m-toluic acid, 4-chloro-3-methyl-benzoic acid, 4-chloro-3-methyl benzoic acid, benzoic acid, 4-chloro-3-methyl, m-toluic acid, 4-chloro, pubchem3666, 5-carboxy-2-chlorotoluene, acmc-1bg77, ksc497q5r, 4-chloranyl-3-methyl-benzoic acid
PubChem CID 282989
IUPAC Name 4-chloro-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1Cl)C(O)=O

Specifications

Melting Point 212°C to 216°C
Quantity 50 g
Beilstein 1936535
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only