missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone

Catalog No. aal0601106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0601106 5 g
AAL0601114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0601106 Supplier Thermo Scientific Chemicals Supplier No. L0601106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37074-39-8 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37074-39-8
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045172
InChI Key XOIGZLJCLDWTQH-UHFFFAOYSA-N
Synonym 1-4-chloro-3-methylphenyl ethanone, 4'-chloro-3'-methylacetophenone, 4-chloro-3-methylacetophenone, ethanone, 1-4-chloro-3-methylphenyl, 1-4-chloro-3-methylphenyl ethan-1-one, 4-chloro-3-methyl acetophenone, 1-4-chloro-3-methyl-phenyl-ethanone, ksc577m5h, 3-methyl-4-chloroacetophenone, 2-methyl-4-acetyl-chlorbenzol
PubChem CID 592743
IUPAC Name 1-(4-chloro-3-methylphenyl)ethanone
SMILES CC(=O)C1=CC(C)=C(Cl)C=C1

Specifications

Density 1.15
Boiling Point 104°C to 105°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 774639
Formula Weight 168.62
Percent Purity 75%
Grade Technical
Assay remainder 3'-chloro-4'-methylacetophenone
Chemical Name or Material 4'-Chloro-3'-methylacetophenone
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only