CAS RN 37074-39-8
IUPAC Name:
1-(4-chloro-3-methylphenyl)ethan-1-one
Synonyms:
1-(4-chloro-3-methylphenyl)ethanone
Molecular Weight (g/mol):
168.62
Molecular Formula:
C9H9ClO
InChi Key:
XOIGZLJCLDWTQH-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(C)=C(Cl)C=C1
1
–
1
of
1
results
1
4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone
CAS: 37074-39-8 Molecular Formula: C9H9ClO Molecular Weight (g/mol): 168.62 MDL Number: MFCD00045172 InChI Key: XOIGZLJCLDWTQH-UHFFFAOYSA-N Synonym: 1-4-chloro-3-methylphenyl ethanone,4'-chloro-3'-methylacetophenone,4-chloro-3-methylacetophenone,ethanone, 1-4-chloro-3-methylphenyl,1-4-chloro-3-methylphenyl ethan-1-one,4-chloro-3-methyl acetophenone,1-4-chloro-3-methyl-phenyl-ethanone,ksc577m5h,3-methyl-4-chloroacetophenone,2-methyl-4-acetyl-chlorbenzol PubChem CID: 592743 IUPAC Name: 1-(4-chloro-3-methylphenyl)ethanone SMILES: CC(=O)C1=CC(C)=C(Cl)C=C1