missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(hydroxymethyl)pyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAH66522MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66522MD 250 mg
AAH6652203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66522MD Supplier Thermo Scientific Chemicals Supplier No. H66522MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 245124-17-8
Molecular Formula C6H7Cl2NO
Molecular Weight (g/mol) 180.028
MDL Number MFCD23703142
InChI Key OPXGRFCMPCHNIY-UHFFFAOYSA-N
Synonym 4-chloropyridin-3-yl methanol hydrochloride, 4-chloropyridin-3-yl methanol hcl, 4-chloro-3-pyridinemethanol hydrochloride, 4-chloro-3-pyridyl methanol hydrochloride, 4-chloropyridin-3-yl methanol-hydrogen chloride 1/1
PubChem CID 71607202
IUPAC Name (4-chloropyridin-3-yl)methanol;hydrochloride
SMILES C1=CN=CC(=C1Cl)CO.Cl

Specifications

Quantity 250 mg
Beilstein 8339779
Formula Weight 180.03
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-3-(hydroxymethyl)pyridine hydrochloride
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only