accessibility menu, dialog, popup

UserName

NClHOHCl

CAS RN 245124-17-8

IUPAC Name: (4-chloropyridin-3-yl)methanol hydrochloride
Synonyms: (4-chloropyridin-3-yl)methanol hydrochloride
Molecular Weight (g/mol): 180.03
Molecular Formula: C6H7Cl2NO
InChi Key: OPXGRFCMPCHNIY-UHFFFAOYSA-N
SMILES: Cl.OCC1=C(Cl)C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results