missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-fluoropyridine 95.0+%, TCI America™
Supplier: TCI America C32451G
Specifications
| 4-Chloro-3-fluoropyridine | |
| -24°C | |
| 139°C | |
| MFCD03453233 | |
| 1993 | |
| BEQUUSCRAKEKQM-UHFFFAOYSA-N | |
| 4-chloro-3-fluoropyridine | |
| 3852429 | |
| ≥95.0% (GC) |
| 2546-56-7 | |
| Yellow | |
| C5H3ClFN | |
| 1 g | |
| pyridine, 4-chloro-3-fluoro, 3-fluoro-4-chloro pyridine, 3-fluoro-4-chloropyridine hydrochloride, pubchem6667, 4-chloro-3fluoropyridine, 3-fluoro-4-chloropyridine, acmc-209x6p, 4-chloro-3-fluoro-pyridin, 3-fluoro-4-chloro-pyridine, 4-chloro-3-fluoro-pyridine | |
| FC1=C(Cl)C=CN=C1 | |
| 131.53 | |
| 131.53 | |
| Liquid |
Chemical Identifiers
| 2546-56-7 | |
| 131.53 | |
| BEQUUSCRAKEKQM-UHFFFAOYSA-N | |
| 3852429 | |
| FC1=C(Cl)C=CN=C1 |
| C5H3ClFN | |
| MFCD03453233 | |
| pyridine, 4-chloro-3-fluoro, 3-fluoro-4-chloro pyridine, 3-fluoro-4-chloropyridine hydrochloride, pubchem6667, 4-chloro-3fluoropyridine, 3-fluoro-4-chloropyridine, acmc-209x6p, 4-chloro-3-fluoro-pyridin, 3-fluoro-4-chloro-pyridine, 4-chloro-3-fluoro-pyridine | |
| 4-chloro-3-fluoropyridine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator