missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. c27491g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27491G Supplier TCI America Supplier No. C27491G
Only null left

Specifications

Chemical Name or Material 4-Chloro-3-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 137504-86-0
Melting Point 144°C
Color White
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD01319010
Synonym 4-chloro-3-fluorophenyl boronic acid, 4-chloro-3-fluorobenzeneboronic acid, 3-fluoro-4-chlorophenylboronic acid, 4-chloro-3-fluoro-phenyl boronic acid, boronic acid, 4-chloro-3-fluorophenyl, 4-chloro-3-fluorobenzeneboronicacid, pubchem1776, ksc174k0r, acmc-1c195, 4-chloro3-fluorophenylboronic acid
InChI Key CMJQIHGBUKZEHP-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)Cl)F)(O)O
IUPAC Name (4-chloro-3-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.362
PubChem CID 2778656
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less