missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorophenol 98.0+%, TCI America™

Catalog No. p-7125483
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24765G 5 g
C247625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24765G Supplier TCI America Supplier No. C24765G
Only null left

Chemical Identifiers

CAS 348-60-7
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00042583
InChI Key XLHYAEBESNFTCA-UHFFFAOYSA-N
Synonym 3-fluoro-4-chlorophenol, 4-chloro-3-fluoro-phenol, phenol, 4-chloro-3-fluoro, pubchem1493, acmc-1ctvv, 3-fluoro-4-chloro phenol, 3-fluoro-4-chloro-phenol, 3-fluoro-4-chlorophenol;, 4-chlor-3-fluor-phenol;, 4-chloro-3-fluorophenol;
PubChem CID 2724523
IUPAC Name 4-chloro-3-fluorophenol
SMILES OC1=CC=C(Cl)C(F)=C1

Specifications

Melting Point 54°C
Color Yellow
Boiling Point 84°C
Quantity 5 g
Formula Weight 146.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-fluorophenol