missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7035606
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
AAH5332506 5 g
1 options

Catalog No. AAH5332506

Supplier: Thermo Scientific Chemicals H5332506

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137504-86-0
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD01319010
InChI Key CMJQIHGBUKZEHP-UHFFFAOYSA-N
Synonym 4-chloro-3-fluorophenyl boronic acid, 4-chloro-3-fluorobenzeneboronic acid, 3-fluoro-4-chlorophenylboronic acid, 4-chloro-3-fluoro-phenyl boronic acid, boronic acid, 4-chloro-3-fluorophenyl, 4-chloro-3-fluorobenzeneboronicacid, pubchem1776, ksc174k0r, acmc-1c195, 4-chloro3-fluorophenylboronic acid
PubChem CID 2778656
IUPAC Name (4-chloro-3-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)Cl)F)(O)O

Specifications

Melting Point 198°C to 204°C
Quantity 5 g
Formula Weight 174.37
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only