missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzaldehyde 97.0+%, TCI America™

Catalog No. C15065G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15065G 5 g
C15061G 1 g
C150625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C15065G Supplier TCI America Supplier No. C15065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5527-95-7
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00143288
InChI Key AZMDWRPTDCIFRD-UHFFFAOYSA-N
Synonym 3-fluoro-4-chlorobenzaldehyde, 4-chloro-3-fluorobenz-aldehyde, 4-chloro-3-fluoro-benzaldehyde, benzaldehyde, 4-chloro-3-fluoro, pubchem1412, acmc-1amt7, ksc494a8n, 4-chloro-3-fluoro benzaldehyde, azmdwrptdcifrd-uhfffaoysa
PubChem CID 2724985
IUPAC Name 4-chloro-3-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)Cl

Specifications

Melting Point 48°C
Quantity 5 g
Formula Weight 158.56
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-fluorobenzaldehyde
TSCA No
Recommended Storage Refrigerator