missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dinitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. C099410G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C099410G 10 g
C099425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C099410G Supplier TCI America Supplier No. C099410G
Only null left

Chemical Identifiers

CAS 393-75-9
Molecular Formula C7H2ClF3N2O4
Molecular Weight (g/mol) 270.548
MDL Number MFCD00007068
InChI Key HFHAVERNVFNSHL-UHFFFAOYSA-N
Synonym 4-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,3-dinitro-5-trifluoromethyl benzene, 3,5-dinitro-4-chlorobenzotrifluoride, benzene, 2-chloro-1,3-dinitro-5-trifluoromethyl, 1-chloro-2,6-dinitro-4-trifluoromethyl benzene, 1,3-dinitro-2-chloro-5-trifluoromethylbenzene, cdnt, chloralin, 2,6-dinitro-4-trifluoromethylchlorobenzene, trifluoromethyl-3,5-dinitro-4-chlorobenzene
PubChem CID 9809
IUPAC Name 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point 56°C
Color Yellow
Quantity 10 g
UN Number 2811
Formula Weight 270.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3,5-dinitrobenzotrifluoride
RTECSNumber XS9065000
TSCA Yes