missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3,5-dimethylphenol 98.0+%, TCI America™
Supplier: TCI America C0161500G
Specifications
| 4-Chloro-3,5-dimethylphenol | |
| 115°C | |
| C8H9ClO | |
| 500 g | |
| chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson | |
| CC1=CC(=CC(=C1Cl)C)O | |
| 156.609 | |
| CHEBI:34393 | |
| ≥98.0% (GC) |
| 88-04-0 | |
| 246°C | |
| MFCD00002324 | |
| 3077 | |
| OSDLLIBGSJNGJE-UHFFFAOYSA-N | |
| 4-chloro-3,5-dimethylphenol | |
| 2723 | |
| 156.61 | |
| Crystalline Powder |
Chemical Identifiers
| 88-04-0 | |
| 156.609 | |
| OSDLLIBGSJNGJE-UHFFFAOYSA-N | |
| 2723 | |
| 4-chloro-3,5-dimethylphenol |
| C8H9ClO | |
| MFCD00002324 | |
| chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson | |
| CHEBI:34393 | |
| CC1=CC(=CC(=C1Cl)C)O |
Safety and Handling
EINECSNumber : (3)-0727&(3)-0936&(9)-1650
RTECSNumber : ZE6850000
TSCA : Yes