missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dimethylphenol 98.0+%, TCI America™

Catalog No. C016125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C016125G 25 g
C0161500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C016125G Supplier TCI America Supplier No. C016125G

Chemical Identifiers

CAS 88-04-0
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002324
InChI Key OSDLLIBGSJNGJE-UHFFFAOYSA-N
Synonym chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson
PubChem CID 2723
ChEBI CHEBI:34393
IUPAC Name 4-chloro-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Cl)C)O

Specifications

Melting Point 115°C
Boiling Point 246°C
Quantity 25 g
UN Number 3077
Formula Weight 156.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3,5-dimethylphenol
EINECSNumber (3)-0727&(3)-0936&(9)-1650
RTECSNumber ZE6850000
TSCA Yes