missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C26011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26011G Supplier TCI America Supplier No. C26011G

Specifications

Chemical Name or Material 4-Chloro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 313545-41-4
Melting Point 182°C
Color White-Yellow
Molecular Formula C7H5BClF3O2
Quantity 1 g
MDL Number MFCD04973086
Synonym 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethylphenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl-phenylboronic acid, boronic acid, 4-chloro-2-trifluoromethyl phenyl, 4-chloro-2-trifluoromethyl phenylboronicacid, pubchem5158, acmc-20aic7
InChI Key YAPBOBGBYQQYHX-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
IUPAC Name [4-chloro-2-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 224.37
PubChem CID 2763244
Formula Weight 224.37
Physical Form Crystalline Powder
Show More Show Less