missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)aniline, 97%

Catalog No. AAB2175514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2175514 25 g
AAB2175522 100 g
AAB2175536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2175514 Supplier Thermo Scientific Chemicals Supplier No. B2175514

CAS: 445-03-4 | C7H5ClF3N | 195.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 445-03-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00007841
InChI Key CVINWVPRKDIGLL-UHFFFAOYSA-N
Synonym 2-amino-5-chlorobenzotrifluoride, 4-chloro-2-trifluoromethyl aniline, benzenamine, 4-chloro-2-trifluoromethyl, 2-amino-5-chlorobenzo trifluoride, 5-chloro-2-aminobenzotrifluoride, 2-amino-5-chloro-trifluorotoluene, 2-trifluoromethyl-4-chloro-aniline, 2-trifluoromethyl-4-chloroaniline, 4-chloro-alpha,alpha,alpha-trifluoro-o-toluidine, 4-chloro-2-trifluoromethyl-phenylamine
PubChem CID 67961
IUPAC Name 4-chloro-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(Cl)C=C1C(F)(F)F

Specifications

Melting Point 8°C to 9°C
Density 1.386
Boiling Point 66°C to 67°C (3 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.507
Quantity 25 g
UN Number UN2810
Beilstein 2366801
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)aniline
Show More Show Less
Hazard Category H227-H302-H315-H318-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-151-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only