missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-pyridinemethanol 98.0+%, TCI America™

Catalog No. p-7125740
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29501G 1 g
C29505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29501G Supplier TCI America Supplier No. C29501G
Only null left

Chemical Identifiers

CAS 63071-10-3
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD07437885
InChI Key UEAIOHHGRGSGGJ-UHFFFAOYSA-N
Synonym 4-Chloro-2-(hydroxymethyl)pyridine
PubChem CID 2763167
IUPAC Name (4-chloropyridin-2-yl)methanol
SMILES C1=CN=C(C=C1Cl)CO

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 66°C
Quantity 1 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-pyridinemethanol