accessibility menu, dialog, popup

UserName

CAS RN 63071-10-3

NClOH

CAS RN 63071-10-3

IUPAC Name: (4-chloropyridin-2-yl)methanol
Synonyms: (4-chloropyridin-2-yl)methanol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: UEAIOHHGRGSGGJ-UHFFFAOYSA-N
SMILES: OCC1=NC=CC(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results