missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-pyridinemethanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 45°C |
| Color | White-Yellow |
| Boiling Point | 66°C |
| Quantity | 1 g |
| Formula Weight | 143.57 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-2-pyridinemethanol |
Safety and Handling
| TSCA | No |