missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrophenol 98.0+%, TCI America™

Catalog No. p-7124296
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C022825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C022825G Supplier TCI America Supplier No. C022825G

Chemical Identifiers

CAS 89-64-5
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD00007113
InChI Key NWSIFTLPLKCTSX-UHFFFAOYSA-N
Synonym 2-nitro-4-chlorophenol, phenol, 4-chloro-2-nitro, unii-438lq62wnh, 4-chloro-2-nitro-phenol, 4-chloro-6-nitrophenol, acmc-209r2b, 5-chloro-2-hydroxynitrobenzene, 4-chloro-2-nitrophenol
PubChem CID 6980
IUPAC Name 4-chloro-2-nitrophenol
SMILES OC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 88°C
Color Yellow
Quantity 25 g
Formula Weight 173.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-nitrophenol
EINECSNumber (3)-0905
TSCA Yes