Learn More
4-Chloro-2-nitrophenol, 98%, contains up to ca 10% water, Thermo Scientific Chemicals
$65.48 - $515.47
Chemical Identifiers
| CAS | 89-64-5 |
|---|---|
| Molecular Formula | C6H4ClNO3 |
| Molecular Weight (g/mol) | 173.55 |
| MDL Number | MFCD00007113 |
| InChI Key | NWSIFTLPLKCTSX-UHFFFAOYSA-N |
| Synonym | 2-nitro-4-chlorophenol, phenol, 4-chloro-2-nitro, unii-438lq62wnh, 4-chloro-2-nitro-phenol, 4-chloro-6-nitrophenol, acmc-209r2b, 5-chloro-2-hydroxynitrobenzene, 4-chloro-2-nitrophenol |
| PubChem CID | 6980 |
| IUPAC Name | 4-chloro-2-nitrophenol |
| SMILES | OC1=CC=C(Cl)C=C1[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1815014
|
Thermo Scientific Chemicals
A1815014 |
25 g |
Each for $65.48
|
|
|||||
|
AAA1815036
|
Thermo Scientific Chemicals
A1815036 |
500 g |
Each for $515.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89-64-5 | |
| 173.55 | |
| NWSIFTLPLKCTSX-UHFFFAOYSA-N | |
| 6980 | |
| OC1=CC=C(Cl)C=C1[N+]([O-])=O |
| C6H4ClNO3 | |
| MFCD00007113 | |
| 2-nitro-4-chlorophenol, phenol, 4-chloro-2-nitro, unii-438lq62wnh, 4-chloro-2-nitro-phenol, 4-chloro-6-nitrophenol, acmc-209r2b, 5-chloro-2-hydroxynitrobenzene, 4-chloro-2-nitrophenol | |
| 4-chloro-2-nitrophenol |
Specifications
| 89-64-5 | |
| C6H4ClNO3 | |
| 25 g | |
| 2-nitro-4-chlorophenol, phenol, 4-chloro-2-nitro, unii-438lq62wnh, 4-chloro-2-nitro-phenol, 4-chloro-6-nitrophenol, acmc-209r2b, 5-chloro-2-hydroxynitrobenzene, 4-chloro-2-nitrophenol | |
| OC1=CC=C(Cl)C=C1[N+]([O-])=O | |
| 173.55 | |
| 173.56 | |
| 4-Chloro-2-nitrophenol |
| 85°C to 88°C | |
| MFCD00007113 | |
| 2048031 | |
| NWSIFTLPLKCTSX-UHFFFAOYSA-N | |
| 4-chloro-2-nitrophenol | |
| 6980 | |
| 98% | |
| contains up to ≈10% water |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 201-927-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only