missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrophenol, 98%, contains up to ca 10% water, Thermo Scientific Chemicals

Catalog No. AAA1815014
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1815014 25 g
AAA1815036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1815014 Supplier Thermo Scientific Chemicals Supplier No. A1815014

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-64-5
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD00007113
InChI Key NWSIFTLPLKCTSX-UHFFFAOYSA-N
Synonym 2-nitro-4-chlorophenol, phenol, 4-chloro-2-nitro, unii-438lq62wnh, 4-chloro-2-nitro-phenol, 4-chloro-6-nitrophenol, acmc-209r2b, 5-chloro-2-hydroxynitrobenzene, 4-chloro-2-nitrophenol
PubChem CID 6980
IUPAC Name 4-chloro-2-nitrophenol
SMILES OC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 85°C to 88°C
Quantity 25 g
Beilstein 2048031
Formula Weight 173.56
Percent Purity 98%
Assay contains up to ≈10% water
Chemical Name or Material 4-Chloro-2-nitrophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 201-927-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only