missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7124660
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C087925G 25 g
C0879500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C087925G Supplier TCI America Supplier No. C087925G
Only null left

Chemical Identifiers

CAS 6280-88-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.56
MDL Number MFCD00007214
InChI Key JAHIPDTWWVYVRV-UHFFFAOYSA-N
Synonym 4-chloro-2-nitrobenzoic acid, 2-nitro-4-chlorobenzoic acid, 4-chloro-2-nitrobenzoicacid, benzoic acid, 4-chloro-2-nitro, 4-chloro-2-nitro-benzoic acid, acmc-1b6vn, ksc353a2d, 2-carboxy-5-chloronitrobenzene, benzoicacid, 4-chloro-2-nitro, 2-nitro-4-chlorobenzoic acid anion
PubChem CID 80475
IUPAC Name 4-chloro-2-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 145°C
Quantity 25 g
Formula Weight 201.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-nitrobenzoic Acid
EINECSNumber (3)-1502
TSCA No