missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrobenzoic acid, 97%

Catalog No. aaa1287214
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
AAA1287214 25 g
AAA1287222 100 g
2 options

Catalog No. AAA1287214

Supplier: Thermo Scientific Chemicals A1287214

Only null left

CAS: 6280-88-2 | C7H4ClNO4 | 201.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6280-88-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.56
MDL Number MFCD00007214
InChI Key JAHIPDTWWVYVRV-UHFFFAOYSA-N
Synonym 4-chloro-2-nitrobenzoic acid, 2-nitro-4-chlorobenzoic acid, 4-chloro-2-nitrobenzoicacid, benzoic acid, 4-chloro-2-nitro, 4-chloro-2-nitro-benzoic acid, acmc-1b6vn, ksc353a2d, 2-carboxy-5-chloronitrobenzene, benzoicacid, 4-chloro-2-nitro, 2-nitro-4-chlorobenzoic acid anion
PubChem CID 80475
IUPAC Name 4-chloro-2-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 142°C to 146°C
Flash Point >100°C (212°F)
Odor Odorless
Quantity 25 g
Beilstein 2051370
Formula Weight 201.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-483-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only