missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-nitrobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America C096425G
Specifications
| 4-Chloro-2-nitrobenzaldehyde | |
| 68°C | |
| MFCD00056104 | |
| MZPNQUMLOFWSEK-UHFFFAOYSA-N | |
| 4-chloro-2-nitrobenzaldehyde | |
| 79674 | |
| ≥98.0% (GC) |
| 5551-11-1 | |
| C7H4ClNO3 | |
| 25 g | |
| C1=CC(=C(C=C1Cl)[N+](=O)[O-])C=O | |
| 185.56 | |
| 185.56 | |
| Crystalline Powder |
Chemical Identifiers
| 5551-11-1 | |
| 185.56 | |
| MZPNQUMLOFWSEK-UHFFFAOYSA-N | |
| 4-chloro-2-nitrobenzaldehyde |
| C7H4ClNO3 | |
| MFCD00056104 | |
| 79674 | |
| C1=CC(=C(C=C1Cl)[N+](=O)[O-])C=O |
Safety and Handling
TSCA : No