missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitroanisole 98.0+%, TCI America™

Catalog No. C021625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C021625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C021625G Supplier TCI America Supplier No. C021625G
Only null left

Chemical Identifiers

CAS 89-21-4
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.58
MDL Number MFCD00024327
InChI Key OSAYFGJUEOYRHY-UHFFFAOYSA-N
Synonym 4-chloro-2-nitroanisole, benzene, 4-chloro-1-methoxy-2-nitro, 2-nitro-4-chloroanisole, anisole, 4-chloro-2-nitro, labotest-bb ltbb001192, labotest-bb lt03381711, 4-methoxy-3-nitrochlorobenzene, 4-chloro-2-nitro-1-methoxybenzene, 4-chlor-2-nitroanisol, 2-nitryl-4-chloroanisole
PubChem CID 66631
IUPAC Name 4-chloro-1-methoxy-2-nitrobenzene
SMILES COC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 98°C
Color Yellow
Quantity 25 g
Formula Weight 187.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-nitroanisole
EINECSNumber (3)-0910&(3)-0911
TSCA No